CAS RN 1207-15-4
CAS RN 1207-15-4
IUPAC Name:
4,12-dimethyl-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms:
4,12-dimethyl-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol):
212.31
Molecular Formula:
C14H12S
InChi Key:
RRYWCJRYULRSJM-UHFFFAOYSA-N
SMILES:
CC1=CC2=C(SC3=C2C=C(C)C=C3)C=C1
1
–
1
of
1
results
1
2,8-Dimethyldibenzothiophene 97.0+%, TCI America™
CAS: 1207-15-4 Molecular Formula: C14H12S Molecular Weight (g/mol): 212.31 MDL Number: MFCD00191350 InChI Key: RRYWCJRYULRSJM-UHFFFAOYSA-N PubChem CID: 50959 IUPAC Name: 4,12-dimethyl-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene SMILES: CC1=CC2=C(SC3=C2C=C(C)C=C3)C=C1