accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120724-84-7

CH3NNSOOOH3CCH3SOOOCH3Na(E/Z)(E/Z)

CAS RN 120724-84-7

IUPAC Name: sodium 2-{3-[3,3-dimethyl-1-(4-sulfonatobutyl)-2,3-dihydro-1H-indol-2-ylidene]prop-1-en-1-yl}-3,3-dimethyl-1-(4-sulfonatobutyl)-3H-indol-1-ium
Synonyms: sodium 2-{3-[3,3-dimethyl-1-(4-sulfonatobutyl)indol-2-ylidene]prop-1-en-1-yl}-3,3-dimethyl-1-(4-sulfonatobutyl)indol-1-ium
Molecular Weight (g/mol): 622.77
Molecular Formula: C31H39N2NaO6S2
InChi Key: SBQIZSUJAUKSIJ-UHFFFAOYSA-M
SMILES: [Na+].CC1(C)C(=CC=CC2=[N+](CCCCS([O-])(=O)=O)C3=CC=CC=C3C2(C)C)N(CCCCS([O-])(=O)=O)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results