Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,3',3'-Tetramethyl-1,1'-bis(4-sulfobutyl)indocarbocyanine Sodium Salt 98.0+%, TCI America™
SDP

Catalog No. D44645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D44645G Supplier TCI America Supplier No. D44645G

Chemical Identifiers

CAS 120724-84-7
Molecular Formula C31H39N2NaO6S2
Molecular Weight (g/mol) 622.771
MDL Number MFCD16621066
InChI Key SBQIZSUJAUKSIJ-UHFFFAOYSA-M
Synonym 2-[3-[3,3-Dimethyl-1-(4-sulfobutyl)-1,3-dihydroindol-2-ylidene]propenyl]-3,3-dimethyl-1-(4-sulfobutyl)-3H-indolium Hydroxide Inner Salt Sodium Salt, 3-[3,3-Dimethyl-1-(4-sulfonatobutyl)indolin-2-ylidene]-1-[3,3-dimethyl-1-(4-sulfonatobutyl)indolium-2-yl]-
PubChem CID 133556299
IUPAC Name sodium;4-[(2E)-2-[(E)-3-[3,3-dimethyl-1-(4-sulfonatobutyl)indol-1-ium-2-yl]prop-2-enylidene]-3,3-dimethylindol-1-yl]butane-1-sulfonate
SMILES CC1(C2=CC=CC=C2[N+](=C1C=CC=C3C(C4=CC=CC=C4N3CCCCS(=O)(=O)[O-])(C)C)CCCCS(=O)(=O)[O-])C.[Na+]

Specifications

Quantity 5 g
Formula Weight 622.77
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,3′,3′-Tetramethyl-1,1′-bis(4-sulfobutyl)indocarbocyanine Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.