CAS RN 120786-18-7
CAS RN 120786-18-7
IUPAC Name:
1-amino-13-ethylidene-11-methyl-6-azatricyclo[7.3.1.0²,⁷]trideca-2(7),3,10-trien-5-one
Synonyms:
1-amino-13-ethylidene-11-methyl-6-azatricyclo[7.3.1.0²,⁷]trideca-2(7),3,10-trien-5-one
Molecular Weight (g/mol):
242.32
Molecular Formula:
C15H18N2O
InChi Key:
ZRJBHWIHUMBLCN-UHFFFAOYNA-N
SMILES:
CC=C1C2CC3=C(C=CC(=O)N3)C1(N)CC(C)=C2
1
–
1
of
1
results
1
MP Biomedicals, Inc (±)-Huperzine A, >98%, MP Biomedicals™
CAS: 120786-18-7 Molecular Formula: C15H18N2O Molecular Weight (g/mol): 242.322 InChI Key: ZRJBHWIHUMBLCN-HZRLMBICSA-N Synonym: huperzine a PubChem CID: 9794806 SMILES: CC=C1C2CC3=C(C1(CC(=C2)C)N)C=CC(=O)N3