Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Huperzine A, >98%, MP Biomedicals™

Catalog No. ICN15879605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15879605 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15879605 Supplier MP Biomedicals, Inc Supplier No. 0215879605

Chemical Identifiers

CAS 120786-18-7
Molecular Formula C15H18N2O
Molecular Weight (g/mol) 242.322
InChI Key ZRJBHWIHUMBLCN-HZRLMBICSA-N
Synonym huperzine a
PubChem CID 9794806
SMILES CC=C1C2CC3=C(C1(CC(=C2)C)N)C=CC(=O)N3

Specifications

Assay Percent Range >98%
Quantity 5 mg
Formula Weight 242.3
Physical Form Solid
Chemical Name or Material (±)-Huperzine A
Recommended Storage Store at 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.