accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeO

CAS RN 12093-10-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 214.05
Molecular Formula: C11H10FeO
InChi Key: —
SMILES: [Fe].c1cccc1.O=Cc1cccc1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)