Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrocenecarboxaldehyde 97.0+%, TCI America™
SDP

Catalog No. F09985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09985G 5 g
F099825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09985G Supplier TCI America Supplier No. F09985G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12093-10-6
Molecular Formula C11H10FeO
Molecular Weight (g/mol) 214.05
MDL Number MFCD00001429
InChI Key UQTCQJVPLIVCAX-UHFFFAOYNA-N
IUPAC Name (cyclopenta-2,4-dien-1-yl)(2-formylcyclopenta-2,4-dien-1-yl)iron
SMILES [Fe].c1cccc1.O=Cc1cccc1

Specifications

Melting Point 115°C
Quantity 5 g
Formula Weight 214.05
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ferrocenecarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.