accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3CHNCH3O

CAS RN 120934-96-5

IUPAC Name: methyl 4-(2-benzylbenzoyl)-2,5-dimethyl-1H-pyrrole-3-carboxylate
Synonyms: methyl 4-(2-benzylbenzoyl)-2,5-dimethyl-1H-pyrrole-3-carboxylate
Molecular Weight (g/mol): 347.41
Molecular Formula: C22H21NO3
InChi Key: MDMWHKZANMNXTF-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)NC(C)=C1C(=O)C1=CC=CC=C1CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results