accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121-05-1

IUPAC Name: (2-aminoethyl)bis(propan-2-yl)amine
Synonyms: (2-aminoethyl)diisopropylamine
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: CURJNMSGPBXOGK-UHFFFAOYSA-N
SMILES: CC(C)N(CCN)C(C)C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (6)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)