Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diisopropylethylenediamine, 97%

Catalog No. AC448251000
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448251000 100 mL
AC448250250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448251000 Supplier Thermo Scientific Chemicals Supplier No. 448251000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 121-05-1 | C8H20N2 | 144.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-05-1
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
InChI Key CURJNMSGPBXOGK-UHFFFAOYSA-N
Synonym 2-aminoethyldiisopropylamine, n,n-diisopropylethylenediamine, 2-diisopropylamino ethylamine, usaf am-2, 1,2-ethanediamine, n,n-bis 1-methylethyl, n,n-diisopropyl-1,2-ethanediamine, n1,n1-diisopropylethane-1,2-diamine, 2-aminoethyl diisopropylamine, n,n-diisopropyl ethylenediamine, 2-diisopropylaminoethylamine
PubChem CID 8459
IUPAC Name N',N'-di(propan-2-yl)ethane-1,2-diamine
SMILES CC(C)N(CCN)C(C)C

Specifications

Density 0.8300g/mL
Boiling Point 170°C
Flash Point 54°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula [(CH3)2CH]2NCH2CH2NH2
Quantity 100 mL
Specific Gravity 0.83
Solubility Information Solubility in water: miscible
Formula Weight 144.26
Percent Purity 97%
Chemical Name or Material N, N-Diisopropylethylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 204-447-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.