accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121-60-8

IUPAC Name: 4-acetamidobenzene-1-sulfonyl chloride
Synonyms: 4-acetamidobenzenesulfonyl chloride
Molecular Weight (g/mol): 233.67
Molecular Formula: C8H8ClNO3S
InChi Key: GRDXCFKBQWDAJH-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)S(Cl)(=O)=O
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)