Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamidobenzenesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. A007425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A007425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A007425G Supplier TCI America Supplier No. A007425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 121-60-8
Melting Point 146°C
Color White-Yellow
Molecular Formula C8H8ClNO3S
Quantity 25 g
MDL Number MFCD00007442
UN Number 3261
Synonym n-acetylsulfanilyl chloride, 4-acetamidobenzene-1-sulfonyl chloride, dagenan chloride, 4-acetylamino benzenesulfonyl chloride, 4-acetamidophenylsulfonyl chloride, benzenesulfonyl chloride, 4-acetylamino, acetylsulfanilyl chloride, p-acetamidobenzenesulfonyl chloride, n-acetylsulphanilyl chloride, p-acetaminobenzenesulfonyl chloride
InChI Key GRDXCFKBQWDAJH-UHFFFAOYSA-N
SMILES CC(=O)NC1=CC=C(C=C1)S(Cl)(=O)=O
IUPAC Name 4-acetamidobenzene-1-sulfonyl chloride
Molecular Weight (g/mol) 233.67
PubChem CID 8481
Formula Weight 233.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamidobenzenesulfonyl Chloride
Show More Show Less
EINECSNumber (3)-1988
RTECSNumber DB8837500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.