accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121-98-2

IUPAC Name: methyl 4-methoxybenzoate
Synonyms: methyl anisate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: DDIZAANNODHTRB-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC)C=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)