accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1210-33-9

IUPAC Name: 2-chlorotricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene
Synonyms: 2-chlorotricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene
Molecular Weight (g/mol): 228.72
Molecular Formula: C15H13Cl
InChi Key: QPERNSDCEUTOTE-UHFFFAOYSA-N
SMILES: ClC1C2=CC=CC=C2CCC2=CC=CC=C12
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)