Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzosuberyl chloride, 98%, Thermo Scientific™

Catalog No. AAL0535506
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0535506 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0535506 Supplier Thermo Scientific Chemicals Supplier No. L0535506

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1210-33-9
Molecular Formula C15H13Cl
Quantity 5g
MDL Number MFCD00003592
UN Number UN3261
Beilstein 612280
Synonym 5-chloro-10,11-dihydro-5h-dibenzo a,d cycloheptene, 5-chlorodibenzosuberane, dibenzosuberyl chloride, 5-chloro-10,11-dihydro-5h-dibenzo a,d 7 annulene, 5h-dibenzo a,d cycloheptene, 5-chloro-10,11-dihydro, 5h-dibenzo a,d cycloheptene, 10,11-dihydro-5-chloro, 2-chlorotricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3,5,7,11,13-hexaene, 2-chlorotricyclo 9.4.0.0 3 ,? pentadeca-1 11 ,3,5,7,12,14-hexaene, dibenzosuberylchloride
InChI Key QPERNSDCEUTOTE-UHFFFAOYSA-N
SMILES ClC1C2=CC=CC=C2CCC2=CC=CC=C12
IUPAC Name 2-chlorotricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene
Molecular Weight (g/mol) 228.72
PubChem CID 14587
Formula Weight 228.72
Percent Purity 98%
Chemical Name or Material Dibenzosuberyl chloride
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECSNumber HO8302000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.