accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121040-08-2

IUPAC Name: 4-[2-(4-pentylcyclohexyl)ethyl]cyclohexan-1-one
Synonyms: 4-[2-(4-pentylcyclohexyl)ethyl]cyclohexan-1-one
Molecular Weight (g/mol): 278.48
Molecular Formula: C19H34O
InChi Key: ISFMATRVDNBZMP-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CCC2CCC(=O)CC2)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results