Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[2-(trans-4-Pentylcyclohexyl)ethyl]cyclohexanone 98.0+%, TCI America™
SDP

Catalog No. P23701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23701G Supplier TCI America Supplier No. P23701G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 121040-08-2
Melting Point 44°C
Color White
Molecular Formula C19H34O
Quantity 1 g
MDL Number MFCD14636435
Synonym 4-[2-(trans-4-Amylcyclohexyl)ethyl]cyclohexanone
InChI Key ISFMATRVDNBZMP-UHFFFAOYSA-N
SMILES CCCCCC1CCC(CC1)CCC2CCC(=O)CC2
IUPAC Name 4-[2-(4-pentylcyclohexyl)ethyl]cyclohexan-1-one
Molecular Weight (g/mol) 278.48
PubChem CID 12030409
Formula Weight 278.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-[2-(trans-4-Pentylcyclohexyl)ethyl]cyclohexanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.