accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH

CAS RN 121193-91-7

IUPAC Name: 2-(naphthalen-1-yl)pyrrolidine
Synonyms: 2-(naphthalen-1-yl)pyrrolidine
Molecular Weight (g/mol): 197.28
Molecular Formula: C14H15N
InChi Key: FEJHFYHZVJWOOE-UHFFFAOYNA-N
SMILES: C1CNC(C1)C1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results