Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Naphthyl)pyrrolidine, 95%, Thermo Scientific™

Catalog No. AAH61800MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61800MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH61800MD Supplier Thermo Scientific Chemicals Supplier No. H61800MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-(1-Naphthyl)pyrrolidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121193-91-7
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD02663484
InChI Key FEJHFYHZVJWOOE-UHFFFAOYSA-N
Synonym 2-naphthalen-1-yl pyrrolidine, 2-naphthalen-1-yl-pyrrolidine, 2-1-naphthyl pyrrolidine, pyrrolidine,2-1-naphthalenyl, acmc-20ct66
PubChem CID 249758
IUPAC Name 2-naphthalen-1-ylpyrrolidine
SMILES C1CC(NC1)C2=CC=CC3=CC=CC=C32

Specifications

Color Yellow
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 197.28
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-(1-Naphthyl)pyrrolidine
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.