accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121219-08-7

IUPAC Name: [4-(hexyloxy)phenyl]boronic acid
Synonyms: 4-(hexyloxy)phenylboronic acid
Molecular Weight (g/mol): 222.09
Molecular Formula: C12H19BO3
InChi Key: XYNVLFGOOWUKQS-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results