Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexyloxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. H13575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1357-5G 5 g
H13571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1357-5G Supplier TCI America Supplier No. H13575G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 121219-08-7
Molecular Formula C12H19BO3
MDL Number MFCD04039027
InChI Key XYNVLFGOOWUKQS-UHFFFAOYSA-N
PubChem CID 3682148

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 5 g
Synonym 4-hexyloxyphenylboronic acid, 4-hexoxyphenyl boronic acid, 4-n-hexyloxy benzeneboronic acid, 4-hexyloxybenzeneboronic acid, 4-hexyloxy phenylboronic acid
SMILES CCCCCCOC1=CC=C(C=C1)B(O)O
IUPAC Name [4-(hexyloxy)phenyl]boronic acid
Molecular Weight (g/mol) 222.09
Formula Weight 222.09
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.