accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBOHOH

CAS RN 121219-12-3

IUPAC Name: (4-pentylphenyl)boronic acid
Synonyms: 4-pentylphenylboronic acid
Molecular Weight (g/mol): 192.07
Molecular Formula: C11H17BO2
InChi Key: UZRMPSOGFATLJE-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results