Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. A23365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23365G Supplier TCI America Supplier No. A23365G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 121219-12-3
Molecular Formula C11H17BO2
Molecular Weight (g/mol) 192.07
MDL Number MFCD00995151
InChI Key UZRMPSOGFATLJE-UHFFFAOYSA-N
Synonym 4-n-pentylphenylboronic acid, 4-pentylphenyl boronic acid, 4-pentylbenzeneboronic acid, 4-n-pentylbenzeneboronic acid, 4-amylphenylboronic acid, 4-pentylbenzene boronic acid, 4-pentylphenylboronicacid, 4-amylbenzeneboronic acid, boronic acid, 4-pentylphenyl, pubchem16788
PubChem CID 4589191
IUPAC Name (4-pentylphenyl)boronic acid
SMILES CCCCCC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 84°C
Color White
Quantity 5 g
Formula Weight 192.07
Physical Form Crystalline Powder
Chemical Name or Material 4-Amylphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.