accessibility menu, dialog, popup

UserName

Viewing profile as user:
PdS

CAS RN 12125-22-3

IUPAC Name: palladium(2+) sulfanediide
Synonyms: palladium(2+) sulfanediide
Molecular Weight (g/mol): 138.48
Molecular Formula: PdS
InChi Key: FEWBURNPAYHODO-UHFFFAOYSA-N
SMILES: [S--].[Pd++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results