Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium Sulfide, MP Biomedicals

Catalog No. ICN205491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205491 Supplier MP Biomedicals, Inc Supplier No. 0520549101

Chemical Identifiers

CAS 12125-22-3
Molecular Formula PdS
Molecular Weight (g/mol) 138.48
MDL Number MFCD00016285
InChI Key FEWBURNPAYHODO-UHFFFAOYSA-N
Synonym palladium ii sulfide, palladium sulphide, palladium monosulphide, palladium sulfide, acmc-20ajzj, palladium sulfide,, palladium ii sulfide trace metals basis
PubChem CID 82926
IUPAC Name palladium(2+) sulfanediide
SMILES [S--].[Pd++]

Specifications

Quantity 1 g
Formula Weight 138.46
Chemical Name or Material Palladium Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.