CAS RN 12125-28-9
IUPAC Name:
magnesium(2+) hydroxide carbonate
Synonyms:
magnesium carbonate hydroxide
Molecular Weight (g/mol):
101.32
Molecular Formula:
CHMgO4
InChi Key:
OUHCLAKJJGMPSW-UHFFFAOYSA-K
SMILES:
[OH-].[Mg++].[O-]C([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Production Chemicals and Services Magnesium Carbonate USP NF FCC, DPL
CAS: 12125-28-9 Molecular Weight (g/mol): 485 g/mol