Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Carbonate USP NF FCC, DPL

Catalog No. 502985629
Encompass
Catalog No. 50-298-5629 Supplier Thermo Scientific Production Chemicals and Services Supplier No. D503036001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12125-28-9
Molecular Weight (g/mol) 485 g/mol

Specifications

Physical Form Powder
Quantity 25 kg
Percent Purity 40 to 43.5%
Grade USP
Packaging 25 kg polylined paperbag
Chemical Name or Material Magnesium carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.