accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121254-39-5

IUPAC Name: {1H-cyclopenta[l]phenanthren-2-yl}methane
Synonyms: 1H-cyclopenta[l]phenanthren-2-ylmethane
Molecular Weight (g/mol): 230.31
Molecular Formula: C18H14
InChi Key: YODBCXSJSTXEIB-UHFFFAOYSA-N
SMILES: CC1=CC2=C(C1)C1=CC=CC=C1C1=CC=CC=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results