Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylcyclopenta[l]phenanthrene 97.0+%, TCI America™
SDP

Catalog No. M14111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14111G 1 g
M1411100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14111G Supplier TCI America Supplier No. M14111G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 121254-39-5
Molecular Formula C18H14
Molecular Weight (g/mol) 230.31
MDL Number MFCD03844801
InChI Key YODBCXSJSTXEIB-UHFFFAOYSA-N
PubChem CID 15842243
IUPAC Name 2-methyl-1H-cyclopenta[l]phenanthrene
SMILES CC1=CC2=C(C1)C3=CC=CC=C3C4=CC=CC=C42

Specifications

Melting Point 166°C
Quantity 1 g
Formula Weight 230.31
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylcyclopenta[l]phenanthrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.