accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121288-39-9

NH2NOHNN(R)O(R)OH(S)OH(R)OHONH2C

CAS RN 121288-39-9

IUPAC Name: 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-7-(prop-2-en-1-yl)-6,7,8,9-tetrahydro-3H-purine-6,8-dione
Synonyms: 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-7-(prop-2-en-1-yl)-3H-purine-6,8-dione
Molecular Weight (g/mol): 339.31
Molecular Formula: C13H17N5O6
InChi Key: VDCRFBBZFHHYGT-IOSLPCCCSA-N
SMILES: NC1=NC(=O)C2=C(N1)N([C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O)C(=O)N2CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results