accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2CHCH2CHClClPdPd

CAS RN 12131-44-1

IUPAC Name: dipalladium(2+) bis(1-phenylprop-2-en-1-ide) dichloride
Synonyms: dipalladium(2+) bis(1-phenylprop-2-en-1-ide) dichloride
Molecular Weight (g/mol): 518.08
Molecular Formula: C18H18Cl2Pd2
InChi Key: PHDNDSSMEIRHSI-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pd++].[Pd++].C=C[CH-]C1=CC=CC=C1.C=C[CH-]C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)