Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II)(pi-cinnamyl) Chloride Dimer 97.0+%, TCI America™
SDP

Catalog No. P2017200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2017200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2017200MG Supplier TCI America Supplier No. P2017200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12131-44-1
Molecular Formula C18H16Cl2Pd2-2
Molecular Weight (g/mol) 516.066
MDL Number MFCD16621470
InChI Key GVZDYJIFRIJSQP-UHFFFAOYSA-L
PubChem CID 131674890
IUPAC Name palladium;prop-2-enylbenzene;dichloride
SMILES [CH+]=C[CH-]C1=CC=CC=C1.[CH+]=C[CH-]C1=CC=CC=C1.[Cl-].[Cl-].[Pd].[Pd]

Specifications

Melting Point 220°C
Quantity 200 mg
Formula Weight 518.08
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Palladium(II)(pi-cinnamyl) Chloride Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.