accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOSOOCl

CAS RN 121315-24-0

IUPAC Name: 4-(2-chloroethanesulfonyl)butanoic acid
Synonyms: 4-(2-chloroethanesulfonyl)butanoic acid
Molecular Weight (g/mol): 214.66
Molecular Formula: C6H11ClO4S
InChi Key: XOPHFHGOBYWODX-UHFFFAOYSA-N
SMILES: OC(=O)CCCS(=O)(=O)CCCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results