Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Chloroethylsulfonyl)butyric Acid 97.0+%, TCI America™
SDP

Catalog No. C24285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24285G Supplier TCI America Supplier No. C24285G

Chemical Identifiers

CAS 121315-24-0
Molecular Formula C6H11ClO4S
Molecular Weight (g/mol) 214.66
InChI Key XOPHFHGOBYWODX-UHFFFAOYSA-N
PubChem CID 18981220
IUPAC Name 4-(2-chloroethylsulfonyl)butanoic acid
SMILES C(CC(=O)O)CS(=O)(=O)CCCl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 214.66
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Chloroethylsulfonyl)butyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.