accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CH3CCH3CH3COCOMo

CAS RN 12132-04-6

IUPAC Name: 1,2,3,4,5-pentamethylcyclopenta-1,3-diene bis(methanidylidyneoxidanium) molybdenum
Synonyms: bis(carbon monoxide) pentamethylcyclopentadiene molybdenum
Molecular Weight (g/mol): 288.21
Molecular Formula: C12H16MoO2
InChi Key: CIKWUWWMMYQFKN-UHFFFAOYSA-N
SMILES: [Mo].[C-]#[O+].[C-]#[O+].CC1C(C)=C(C)C(C)=C1C
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)