accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12136-78-6

IUPAC Name: (silanylidenemolybdenio)silane
Synonyms: molybdenum disilicide
Molecular Weight (g/mol): 156.15
Molecular Formula: H4MoSi2
InChi Key: REHMQSAMQCFKDZ-UHFFFAOYSA-N
SMILES: [SiH2]=[Mo]=[SiH2]
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)