Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum silicide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis), Thermo Scientific™

Catalog No. AA41077KS
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41077KS 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41077KS Supplier Thermo Scientific Chemicals Supplier No. 41077KS

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12136-78-6
Molecular Formula MoSi2
Molecular Weight (g/mol) 152.12
MDL Number MFCD00151535
InChI Key YXTPWUNVHCYOSP-UHFFFAOYSA-N
Synonym molybdenum disilicide, molybdenum silicide, ms silicide, molybdenum-silicide, molybdenum silicide, mo5si3, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 6.35mm 0.250in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 6.35mm 0.250in thick
PubChem CID 6336985
IUPAC Name bis($l^{2}-silanylidene)molybdenum
SMILES [Si]=[Mo]=[Si]

Specifications

Color Gray
Melting Point 1870°C to 2030°C
Density 6.31 g/mL
Quantity 1 Ea.
Odor Odorless
Formula Weight 152.13
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Molybdenum silicide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
EINECSNumber 234-786-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.