accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1214319-92-2

CAS RN 1214319-92-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results