CAS RN 1214384-93-6
IUPAC Name:
3-methoxy-2-(trifluoromethyl)benzoic acid
Synonyms:
3-methoxy-2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol):
220.15
Molecular Formula:
C9H7F3O3
InChi Key:
UOBKAUNYVVOXMY-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(C(O)=O)=C1C(F)(F)F
1
–
1
of
1
results
1
3-Methoxy-2-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™
CAS: 1214384-93-6 Molecular Formula: C9H7F3O3 Molecular Weight (g/mol): 220.147 MDL Number: MFCD13185681 InChI Key: UOBKAUNYVVOXMY-UHFFFAOYSA-N Synonym: 3-methoxy-2-trifluoromethyl benzoic acid PubChem CID: 73012640 IUPAC Name: 3-methoxy-2-(trifluoromethyl)benzoic acid SMILES: COC1=CC=CC(=C1C(F)(F)F)C(=O)O