accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12145-90-3

IUPAC Name: bis(chloromercury) bis(cyclopenta-1,3-diene-1,2,3,4,5-pentaide) iron
Synonyms: bis(cyclopenta-1,3-diene-1,2,3,4,5-pentaide) bis(mercury chloride) iron
Molecular Weight (g/mol): 650.06
Molecular Formula: C10H2Cl2FeHg2
InChi Key: RFGKJRRCOWYEHL-UHFFFAOYSA-L
SMILES: [Fe].Cl[Hg].Cl[Hg].[CH-]1[C-]=[C-][C-]=[C-]1.[CH-]1[C-]=[C-][C-]=[C-]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results