Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis(chloromercuri)ferrocene (Tech.), Thermo Scientific™

Catalog No. AC168920010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC168920010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC168920010 Supplier Thermo Scientific Chemicals Supplier No. 168920010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12145-90-3
Molecular Formula C10H2Cl2FeHg2-10
Molecular Weight (g/mol) 650.055
InChI Key RFGKJRRCOWYEHL-UHFFFAOYSA-L
Synonym 1,1'-bis chloromercuri ferrocene
PubChem CID 5148315
IUPAC Name chloromercury;cyclopenta-1,3-diene;iron
SMILES [CH-]1[C-]=[C-][C-]=[C-]1.[CH-]1[C-]=[C-][C-]=[C-]1.Cl[Hg].Cl[Hg].[Fe]

Specifications

Quantity 1 g
Formula Weight 656.1
Chemical Name or Material 1, 1'-Bis(chloromercuri)ferrocene, tech.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.