accessibility menu, dialog, popup

UserName

Viewing profile as user:
SiCuCuCuCuCu

CAS RN 12159-07-8

IUPAC Name: pentacopper silicon
Synonyms: copper silicide
Molecular Weight (g/mol): 345.82
Molecular Formula: Cu5Si
InChi Key: JUZTWRXHHZRLED-UHFFFAOYSA-N
SMILES: [Si].[Cu].[Cu].[Cu].[Cu].[Cu]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results