Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper silicide, 99.5% (metals basis)

Catalog No. AA1284422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1284422 100 g
AA1284414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1284422 Supplier Thermo Scientific Chemicals Supplier No. 01284422
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12159-07-8 | Cu5Si

Copper silicide thin film is used for passivation of copper-based chips, where it serves to suppress diffusion and electromigration and serves as a diffusion barrier. It is involved in the direct process, the industrial route to organosilicon compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12159-07-8
Molecular Formula Cu5Si
MDL Number MFCD00134719

Specifications

Melting Point 825°C
Color Silver
Physical Form Pieces
Quantity 100 g
Odor Odorless
Assay Percent Range 99.5% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 345.79
Chemical Name or Material Copper silicide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-286-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.