accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12166-20-0

SRuS

CAS RN 12166-20-0

IUPAC Name: disulfanylideneruthenium
Synonyms: ruthenium(IV) sulfide
Molecular Weight (g/mol): 165.19
Molecular Formula: RuS2
InChi Key: XNQLQKWNBPVVGB-UHFFFAOYSA-N
SMILES: S=[Ru]=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results