accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNONOHHCl

CAS RN 1216621-00-9

IUPAC Name: 1-[1-(cyclooctylmethyl)-5-(hydroxymethyl)-1,2,3,6-tetrahydropyridin-4-yl]-3-ethyl-2,3-dihydro-1H-1,3-benzodiazol-2-one hydrochloride
Synonyms: 1-[1-(cyclooctylmethyl)-3-(hydroxymethyl)-5,6-dihydro-2H-pyridin-4-yl]-3-ethyl-1,3-benzodiazol-2-one hydrochloride
Molecular Weight (g/mol): 434.02
Molecular Formula: C24H36ClN3O2
InChi Key: YEVMLMQAMCKVSC-UHFFFAOYSA-N
SMILES: Cl.CCN1C(=O)N(C2=CC=CC=C12)C1=C(CO)CN(CC2CCCCCCC2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results