accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1217-45-4

IUPAC Name: anthracene-9,10-dicarbonitrile
Synonyms: 9,10-dicyanoanthracene
Molecular Weight (g/mol): 228.25
Molecular Formula: C16H8N2
InChi Key: BIOPPFDHKHWJIA-UHFFFAOYSA-N
SMILES: N#CC1=C2C=CC=CC2=C(C#N)C2=CC=CC=C12
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results