accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONO

CAS RN 1217-89-6

IUPAC Name: 1-benzyl-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 1-benzylindole-2,3-dione
Molecular Weight (g/mol): 237.26
Molecular Formula: C15H11NO2
InChi Key: SIISFRLGYDVIRG-UHFFFAOYSA-N
SMILES: O=C1N(CC2=CC=CC=C2)C2=CC=CC=C2C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results