Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzylisatin 97.0+%, TCI America™
SDP

Catalog No. B47511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47511G Supplier TCI America Supplier No. B47511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1217-89-6
Molecular Formula C15H11NO2
Molecular Weight (g/mol) 237.26
MDL Number MFCD00187663
InChI Key SIISFRLGYDVIRG-UHFFFAOYSA-N
Synonym 1-Benzylindole-2,3-dione, 1-Benzyl-2,3-indolinedione
PubChem CID 264734
IUPAC Name 1-benzyl-2,3-dihydro-1H-indole-2,3-dione
SMILES O=C1N(CC2=CC=CC=C2)C2=CC=CC=C2C1=O

Specifications

Melting Point 130°C
Quantity 1 g
Formula Weight 237.26
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzylisatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.