accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3HNOSClHCl(E/Z)

CAS RN 1217201-17-6

IUPAC Name: 3-{2-[(4-chlorophenyl)sulfanyl]phenyl}-N-[4-(dimethylamino)butyl]prop-2-enamide hydrochloride
Synonyms: 3-{2-[(4-chlorophenyl)sulfanyl]phenyl}-N-[4-(dimethylamino)butyl]prop-2-enamide hydrochloride
Molecular Weight (g/mol): 425.41
Molecular Formula: C21H26Cl2N2OS
InChi Key: PDVBHWZPRQFKJS-UHFFFAOYSA-N
SMILES: Cl.CN(C)CCCCNC(=O)C=CC1=CC=CC=C1SC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results