accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNNHONONH2NCH3H3CFFF

CAS RN 1217486-61-7

IUPAC Name: N1-{4-methyl-5-[2-(1,1,1-trifluoro-2-methylpropan-2-yl)pyridin-4-yl]-1,3-thiazol-2-yl}pyrrolidine-1,2-dicarboxamide
Synonyms: N1-{4-methyl-5-[2-(1,1,1-trifluoro-2-methylpropan-2-yl)pyridin-4-yl]-1,3-thiazol-2-yl}pyrrolidine-1,2-dicarboxamide
Molecular Weight (g/mol): 441.47
Molecular Formula: C19H22F3N5O2S
InChi Key: STUWGJZDJHPWGZ-UHFFFAOYNA-N
SMILES: CC1=C(SC(NC(=O)N2CCCC2C(N)=O)=N1)C1=CC(=NC=C1)C(C)(C)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)